4-Chloro-o-phenylenediamine ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Chloro-3,4-diaminobenzene, 4-Chlorobenzene-1,2-diamine, 4-Chloro-1,2-phenylenediamine, 4-Chloro-1,2-diaminobenzene
075185-100G
075185-25G
102540-200EA
255.38
USD
102540-200
102664-036
4-Chloro-o-phenylenediamine ≥95%
4-Chloro-o-phenylenediamine
Hazard: Irritant-Harmful
Formula:
C₆H₇ClN₂ MW: 142.59 g/mol Melting Pt: 70-74°C |
MDL Number:
MFCD00011691 CAS Number: 95-83-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...