4-Decylaniline ≥95%

Supplier: Matrix Scientific

075184-1G 075184-5G 075184-10G
102540-190EA 72.31 USD
102540-190 102540-192 102540-194
4-Decylaniline ≥95%
4-Decylaniline

Hazard: Irritant/Air Sensitive

Formula: C₁₆H₂₇N
MW: 233.4 g/mol
Boiling Pt: 218…220 °C (14 mmHg)
Melting Pt: 23…25 °C
Flash Pt: >110 °C
MDL Number: MFCD00007918
CAS Number: 37529-30-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR