(R)-Piperazine-2-carboxylic acid dihydrochloride ≥95%

Supplier: Matrix Scientific

075180-1G 075180-5G
102540-172EA 21.54 USD
102540-172 102540-174
(R)-Piperazine-2-carboxylic acid dihydrochloride ≥95%
(R)-Piperazine-2-carboxylic acid dihydrochloride

Hazard: Irritant

Formula: C₅H₁₂Cl₂N₂O₂
MDL Number: MFCD03840829
CAS Number: 126330-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR