cis-4-Hydroxy-D(+)-proline ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-(+)-cis-4-Hydroxyproline, cis-4-Hydroxy-D-proline, H-D-cis-Hyp-OH, H-cis-D-Hyp-OH, D(+)-cis-4-Hydroxyproline

075178-5G 075178-100G
102540-160EA 73.85 USD
102540-160 102540-164
cis-4-Hydroxy-D(+)-proline ≥95%
cis-4-Hydroxy-D(+)-proline

Hazard: Irritant

Formula: C₅H₉NO₃
MW: 131.13 g/mol
Melting Pt: 243 °C. (dec.)
MDL Number: MFCD00005252
CAS Number: 2584-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR