4-Chloro-3-fluorotoluene ≥95%

Supplier: Matrix Scientific

075175-1G 075175-5G 075175-10G
102540-146EA 72.31 USD
102540-146 102540-148 102540-150
4-Chloro-3-fluorotoluene ≥95%
4-Chloro-3-fluorotoluene

Hazard: Irritant

Formula: C₇H₆ClF
MW: 144.58 g/mol
MDL Number: MFCD00143295
CAS Number: 5527-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR