3-Hydroxy-2-pyridone ≥95%

Supplier: Matrix Scientific

075174-100G 075174-25G
102540-144EA 333.85 USD
102540-144 102833-790
3-Hydroxy-2-pyridone ≥95%
3-Hydroxy-2-pyridone

Hazard: Irritant

Formula: C₅H₅NO₂
MW: 111.1 g/mol
Melting Pt: ca 249°C dec
MDL Number: MFCD00006271
CAS Number: 16867-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR