4-Formyl-N-isopropylbenzamide ≥95%

Supplier: Matrix Scientific

075172-1G 075172-5G
102540-130EA 640 USD
102540-130 102540-132
4-Formyl-N-isopropylbenzamide ≥95%
4-Formyl-N-isopropylbenzamide

Hazard: Irritant

Formula: C₁₁H₁₃NO₂
MW: 191.2300 g/mol
MDL Number: MFCD03840703
CAS Number: 13255-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR