4-Bromomethyl-1,2-dihydroquinoline-2-one ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Bromomethyl)quinolin-2(1H)-one

075171-1G 075171-5G 075171-10G
102540-124EA 44.62 USD
102540-124 102540-126 102540-128
4-Bromomethyl-1,2-dihydroquinoline-2-one ≥95%
4-Bromomethyl-1,2-dihydroquinoline-2-one

Hazard: Irritant

Formula: C₁₀H₈BrNO
MW: 238.08 g/mol
Melting Pt: 256…258 °C
MDL Number: MFCD03426152
CAS Number: 4876-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR