(4-(Benzyloxy)phenyl)hydrazine hydrochloride ≥95%

Supplier: Matrix Scientific

075168-1G 075168-5G 075168-10G
102540-106EA 38.46 USD
102540-106 102540-108 102540-110
(4-(Benzyloxy)phenyl)hydrazine hydrochloride ≥95%
(4-(Benzyloxy)phenyl)hydrazine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₅ClN₂O
MDL Number: MFCD00137741
CAS Number: 52068-30-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR