4-Aminosalicylic acid ≥95%

Supplier: Matrix Scientific

075166-10G 075166-100G
102540-096EA 36.92 USD
102540-096 102540-098
4-Aminosalicylic acid ≥95%
4-Aminosalicylic acid

Hazard: Irritant

Formula: C₇H₇NO₃
MDL Number: MFCD00007789
CAS Number: 65-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR