4-(2-Naphthyl)phenylboronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (4-(Naphthalen-2-yl)phenyl)boronic acid

075165-1G 075165-5G
102540-090EA 106.15 USD
102540-090 102540-092
4-(2-Naphthyl)phenylboronic acid ≥95%
4-(2-Naphthyl)phenylboronic acid

Hazard: Irritant

Formula: C₁₆H₁₃BO₂
MW: 248.0800 g/mol
MDL Number: MFCD09260454
CAS Number: 918655-03-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR