Indole-2-acetic acid ≥95%

Supplier: Matrix Scientific

075163-1G 075163-5G
102540-080EA 512.31 USD
102540-080 102540-082
Indole-2-acetic acid ≥95%
Indole-2-acetic acid

Hazard: Irritant

Formula: C₁₀H₉NO₂
MW: 175.1840 g/mol
MDL Number: MFCD00957069
CAS Number: 32588-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR