4-(1-Pyrrolidinyl)piperidine ≥95%

Supplier: Matrix Scientific

075162-1G 075162-5G 075162-10G
102540-074EA 55.38 USD
102540-074 102540-076 102540-078
4-(1-Pyrrolidinyl)piperidine ≥95%
4-(1-Pyrrolidinyl)piperidine

Hazard: Irritant

Formula: C₉H₁₈N₂
MW: 154.2526 g/mol
Melting Pt: 57…59 °C
MDL Number: MFCD00038011
CAS Number: 5004-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR