trans-4-Cyclohexyl-L-proline hydrochloride ≥95%

Supplier: Matrix Scientific

075161-1G 075161-5G
102540-070EA 98.46 USD
102540-070 102540-072
trans-4-Cyclohexyl-L-proline hydrochloride ≥95%
trans-4-Cyclohexyl-L-proline hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₀ClNO₂
MDL Number: MFCD08704794
CAS Number: 90657-55-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR