3-Methyl-2-cyclopenten-1-one ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Methylcyclopent-2-enone, 3-Methyl-2-cyclopentenone
075160-5G
075160-10G
075160-100G
102540-064EA
61.54
USD
102540-064
102540-066
102540-068
3-Methyl-2-cyclopenten-1-one ≥95%
3-Methyl-2-cyclopenten-1-one
Hazard: Irritant
Formula:
C₆H₈O MW: 96.13 g/mol |
MDL Number:
MFCD00001403 CAS Number: 2758-18-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...