3-Hydroxy-3-phenylpropanoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Hydroxy-3-phenylpropionic acid

075158-1G 075158-5G
102540-056EA 284.62 USD
102540-056 102540-058
3-Hydroxy-3-phenylpropanoic acid ≥95%
3-Hydroxy-3-phenylpropanoic acid

Hazard: Irritant

Formula: C9H10O3
MW: 166.1700 g/mol
MDL Number: MFCD00137907
CAS Number: 3480-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR