2,2-Dimethyl-1-phenylpropylamine ≥95%

Supplier: Matrix Scientific

Synonyms: 2,2-Dimethyl-1-phenylpropan-1-amine

075157-1G 075157-5G
102540-052EA 355.38 USD
102540-052 102540-054
2,2-Dimethyl-1-phenylpropylamine ≥95%
2,2-Dimethyl-1-phenylpropylamine

Hazard: Irritant

Formula: C₁₁H₁₇N
MW: 163.26 g/mol
MDL Number: MFCD00267378
CAS Number: 61501-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR