N-Boc-L-threonine ≥95%

Supplier: Matrix Scientific

Synonyms: N-Boc-L-threonine, Boc-Thr-OH, (2S,3R)-2-((tert-Butoxycarbonyl)amino)-3-hydroxybutanoic acid

075152-10G 075152-100G
102540-030EA 30.77 USD
102540-030 102540-032
N-Boc-L-threonine ≥95%
N-Boc-L-threonine

Hazard: Irritant

Formula: C₉H₁₇NO₅
MW: 219.2 g/mol
MDL Number: MFCD00065946
CAS Number: 2592-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR