5-Bromoisatin ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromoindoline-2,3-dione, 5-Bromo-1H-indole-2,3-dione

075149-5G 075149-10G 075149-100G
102540-012EA 36.92 USD
102540-012 102540-014 102540-016
5-Bromoisatin ≥95%
5-Bromoisatin

Hazard: Irritant

Formula: C₈H₄BrNO₂
MDL Number: MFCD00149345
CAS Number: 87-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR