4-Cyanobenzoic acid ≥95%

Supplier: Matrix Scientific

075148-10G 075148-100G
102540-008EA 1996.92 USD
102540-008 102540-010
4-Cyanobenzoic acid ≥95%
4-Cyanobenzoic acid

Hazard: Irritant

Formula: C₈H₅NO₂
MW: 147.13 g/mol
MDL Number: MFCD00002528
CAS Number: 619-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR