3-Iodophenylboronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: Dihydroxy(3-iodophenyl)borane, (3-Iodophenyl)boronic acid, 3-Iodobenzeneboronic acid, (3-Iodophenyl)metaboric acid

075147-1G 075147-5G 075147-10G
102540-002EA 107.69 USD
102540-002 102540-004 102540-006
3-Iodophenylboronic acid ≥95%
3-Iodophenylboronic acid

Hazard: Irritant

Formula: C₆H₆BIO₂
MW: 247.83 g/mol
MDL Number: MFCD01319015
CAS Number: 221037-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR