3,4-Diaminotoluene ≥95%

Supplier: Matrix Scientific

Synonyms: 1,2-Diamino-4-methylbenzene, Toluene-3,4-diamine

075144-10G 075144-100G
102539-988EA 36.92 USD
102539-988 102539-990
3,4-Diaminotoluene ≥95%
3,4-Diaminotoluene

Hazard: Irritant

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
MDL Number: MFCD00007728
CAS Number: 496-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR