3,4-Dehydro-L-proline ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-2,5-Dihydro-1H-pyrrole-2-carboxylic acid, (S)-3-Pyrroline-2-carboxylic Acid, H-3,4-Dehydro-Pro-OH

075143-250MG 075143-1G 075143-5G
102539-982EA 400 USD
102539-982 102539-984 102539-986
3,4-Dehydro-L-proline ≥95%
3,4-Dehydro-L-proline

Hazard: Irritant

Formula: C₅H₇NO₂
MW: 113.12 g/mol
MDL Number: MFCD00065964
CAS Number: 4043-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR