3,5-Dimethylbenzoic acid ≥95%

Supplier: Matrix Scientific

075141-10G 075141-100G
102539-972EA 36.92 USD
102539-972 102539-974
3,5-Dimethylbenzoic acid ≥95%
3,5-Dimethylbenzoic acid

Hazard: Irritant

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
MDL Number: MFCD00002525
CAS Number: 499-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR