3-(tert-Butyldimethylsiloxy)glutaric anhydride ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(tert-Butyldimethylsilyloxy)glutaric anhydride

075140-1G 075140-5G 075140-10G
102539-966EA 427.69 USD
102539-966 102539-968 102539-970
3-(tert-Butyldimethylsiloxy)glutaric anhydride ≥95%
3-(tert-Butyldimethylsiloxy)glutaric anhydride

Hazard: Irritant

Formula: C₁₁H₂₀O₄Si
MW: 244.362 g/mol
MDL Number: MFCD00075235
CAS Number: 91424-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR