4-Bromomethyl-2-cyanobiphenyl ≥95%
Supplier: Matrix Scientific
Synonyms:
4'-(Bromomethyl)-[1,1'-biphenyl]-2-carbonitrile
075139-5G
075139-10G
075139-100G
102539-960EA
107.69
USD
102539-960
102539-962
102539-964
4-Bromomethyl-2-cyanobiphenyl ≥95%
4-Bromomethyl-2-cyanobiphenyl
Hazard: Irritant
Formula:
C₁₄H₁₀BrN MW: 272.14 g/mol |
MDL Number:
MFCD00671503 CAS Number: 114772-54-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...