3-(4-Cyanophenyl)propanoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(4-Cyanophenyl)propionic acid

075137-1G 075137-5G 075137-10G
102539-948EA 355.38 USD
102539-948 102539-950 102539-952
3-(4-Cyanophenyl)propanoic acid ≥95%
3-(4-Cyanophenyl)propanoic acid

Hazard: Irritant

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
MDL Number: MFCD08234823
CAS Number: 42287-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR