2-Phenylthiazole ≥95%

Supplier: Matrix Scientific

075135-1G 075135-5G
102539-938EA 640 USD
102539-938 102539-940
2-Phenylthiazole ≥95%
2-Phenylthiazole

Hazard: Irritant

Formula: C₉H₇NS
MW: 161.2200 g/mol
MDL Number: MFCD00272327
CAS Number: 1826-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR