4'-Methoxyacetoacetanilide ≥95%

Supplier: Matrix Scientific

075134-5G 075134-10G 075134-100G
102539-932EA 107.69 USD
102539-932 102539-934 102539-936
4'-Methoxyacetoacetanilide ≥95%
4'-Methoxyacetoacetanilide

Hazard: Irritant

Formula: C₁₁H₁₃NO₃
MW: 207.2300 g/mol
MDL Number: MFCD00008783
CAS Number: 5437-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR