3-Nitro-o-toluic acid ≥95%

Supplier: Matrix Scientific

075133-10G 075133-100G
102539-928EA 36.92 USD
102539-928 102539-930
3-Nitro-o-toluic acid ≥95%
3-Nitro-o-toluic acid

Hazard: Irritant

Formula: C₈H₇NO₄
MDL Number: MFCD00007160
CAS Number: 1975-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR