2-Methyl-1-naphthol ≥95%
Supplier: Matrix Scientific
075131-1G
075131-5G
075131-10G
102539-918EA
284.62
USD
102539-918
102539-920
102539-922
2-Methyl-1-naphthol ≥95%
2-Methyl-1-naphthol
Hazard: Irritant
Formula:
C₁₁H₁₀O MW: 158.2 g/mol |
MDL Number:
MFCD00003964 CAS Number: 7469-77-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...