4-Acetylbenzoic acid ≥95%

Supplier: Matrix Scientific

075130-5G 075130-100G
102539-912EA 43.08 USD
102539-912 102539-916
4-Acetylbenzoic acid ≥95%
4-Acetylbenzoic acid

Hazard: Irritant

Formula: C₉H₈O₃
MDL Number: MFCD00002561
CAS Number: 586-89-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR