Methyl-3-formylbenzoate ≥95%

Supplier: Matrix Scientific

075129-5G 075129-25G
102539-908EA 107.69 USD
102539-908 102833-782
Methyl-3-formylbenzoate ≥95%
Methyl-3-formylbenzoate

Hazard: Irritant-Harmful

Formula: C₉H₈O₃
Melting Pt: 48…52 °C
MDL Number: MFCD00189379
CAS Number: 52178-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR