2-Mercaptobenzimidazole ≥95%

Supplier: Matrix Scientific

Synonyms: Benzimidazole-2-thiol, 1,3-Dihydro-2H-benzo[d]imidazole-2-thione, Rabeprazole Related Compound C, 1,3-Dihydro-2H-benzimidazole-2-thione, 2-Benzimidazolethiol

075128-100G 075128-25G
102539-904EA 107.69 USD
102539-904 102833-780
2-Mercaptobenzimidazole ≥95%
2-Mercaptobenzimidazole

Hazard: Irritant/Refrigerate/Stench/Stored under Argon

Formula: C₇H₆N₂S
MW: 150.2 g/mol
Melting Pt: 300-305°C
MDL Number: MFCD00466107
CAS Number: 583-39-1
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR