(S)-2-Hydroxy-2-phenylethyl-4-methylbenzenesulfonate ≥95%
Supplier: Matrix Scientific
Synonyms:
(S)-2-Hydroxy-2-phenylethyl 4-methylbenzenesulphonate
075127-1G
075127-5G
075127-10G
102539-894EA
100
USD
102539-894
102539-896
102539-898
(S)-2-Hydroxy-2-phenylethyl-4-methylbenzenesulfonate ≥95%
(S)-2-Hydroxy-2-phenylethyl-4-methylbenzenesulfonate
Hazard: Irritant
Formula:
C₁₅H₁₆O₄S MW: 292.3501 g/mol |
MDL Number:
MFCD00013428 CAS Number: 40435-14-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...