Boc-L-homoserine ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-2-(Boc-amino)-4-hydroxybutyric acid, Boc-L-Homoserine, (S)-2-(tert-Butoxycarbonylamino)-4-hydroxybutyric acid, N-Boc-L-homoserine, N-(tert-Butoxycarbonyl)-L-homoserine

075122-1G 075122-5G
102539-870EA 36.92 USD
102539-870 102539-872
Boc-L-homoserine ≥95%
Boc-L-homoserine

Hazard: Irritant

Formula: C₉H₁₇NO₅
MW: 219.2350 g/mol
MDL Number: MFCD00057839
CAS Number: 41088-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR