4-Nitro-L-phenylalanine monohydrate ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-2-Amino-3-(4-nitrophenyl)propanoic acid monohydrate, (S)-4-Nitrophenylalanine monohydrate, H-Phe(4-NO2)-OH.H2O

075116-10G 075116-100G
102539-842EA 143.08 USD
102539-842 102539-844
4-Nitro-L-phenylalanine monohydrate ≥95%
4-Nitro-L-phenylalanine monohydrate

Hazard: Irritant

Formula: C₉H₁₂N₂O₅
MW: 228.20 g/mol
MDL Number: MFCD00150543
CAS Number: 207591-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR