1,4-Dithiane-2,5-diol ≥95%
Supplier: Matrix Scientific
Synonyms:
2,5-Dihydroxy-1,4-dithiane, p-Dithiane-2,5-diol
075113-10G
075113-100G
102539-828EA
36.92
USD
102539-828
102539-830
1,4-Dithiane-2,5-diol ≥95%
1,4-Dithiane-2,5-diol
Hazard: Irritant
Formula:
C₄H₈O₂S₂ MW: 152.24 g/mol |
MDL Number:
MFCD00006659 CAS Number: 40018-26-6 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...