1,4-Difluoro-2-nitrobenzene ≥95%

Supplier: Matrix Scientific

Synonyms: 2,5-Difluoronitrobenzene

075112-10G 075112-100G
102539-824EA 55.38 USD
102539-824 102539-826
1,4-Difluoro-2-nitrobenzene ≥95%
1,4-Difluoro-2-nitrobenzene

Hazard: Irritant

Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
Boiling Pt: 206-207°C
Density: 1.467
Flash Pt: 90°C
MDL Number: MFCD00007054
CAS Number: 364-74-9
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR