3-(Indol-3-yl)propionic acid ≥95%

Supplier: Matrix Scientific

075111-100G 075111-25G
102539-822EA 73.85 USD
102539-822 102833-778
3-(Indol-3-yl)propionic acid ≥95%
3-(Indol-3-yl)propionic acid

Hazard: Irritant

Formula: C₁₁H₁₁NO₂
Melting Pt: 132…135 °C
MDL Number: MFCD00005660
CAS Number: 830-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR