(S)-2-Hydroxy-3-phenylpropanoic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
L-β-Phenyllactic acid, (S)-(-)-3-Phenyllactic Acid, L-3-Phenyllactic Acid, L(-)-3-Phenyllactic acid, (S)-(-)-2-Hydroxy-3-phenylpropionic Acid, L-(-)-3-Phenyllactic acid
075109-5G
075109-10G
075109-100G
102539-808EA
46.15
USD
102539-808
102539-810
102539-812
(S)-2-Hydroxy-3-phenylpropanoic acid ≥95%
(S)-2-Hydroxy-3-phenylpropanoic acid
Hazard: Irritant
Formula:
C₉H₁₀O₃ MW: 166.17 g/mol |
MDL Number:
MFCD00004244 CAS Number: 20312-36-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...