2,4-Dibromobenzoic acid ≥95%
Supplier: Matrix Scientific
075108-1G
075108-5G
102539-802EA
16.92
USD
102539-802
102539-804
2,4-Dibromobenzoic acid ≥95%
2,4-Dibromobenzoic acid
Hazard: Irritant
Formula:
C₇H₄Br₂O₂ MW: 279.92 g/mol |
MDL Number:
MFCD00234253 CAS Number: 611-00-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...