4-Hydroxy-3-methylbenzaldehyde ≥95%
Supplier: Matrix Scientific
075104-1G
075104-5G
102539-782EA
72.31
USD
102539-782
102539-784
4-Hydroxy-3-methylbenzaldehyde ≥95%
4-Hydroxy-3-methylbenzaldehyde
Hazard: Irritant
Formula:
C₈H₈O₂ MW: 136.1479 g/mol |
MDL Number:
MFCD00012360 CAS Number: 15174-69-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...