6-(Methoxycarbonyl)picolinic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2,6-Pyridinedicarboxylic acid monomethyl ester, Dipicolinic acid monomethyl ester, Monomethyl 2,6-Pyridinedicarboxylate, Monomethyl dipicolinate

075100-1G 075100-5G
102539-760EA 35.38 USD
102539-760 102539-762
6-(Methoxycarbonyl)picolinic acid ≥95%
6-(Methoxycarbonyl)picolinic acid

Hazard: Irritant

Formula: C8H7NO4
MW: 181.1455 g/mol
MDL Number: MFCD00806109
CAS Number: 7170-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR