Ethyl-2-((2-((tert-butoxycarbonyl)amino)ethyl)amino)acetate hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-((2-((tert-butoxycarbonyl)amino)ethyl)amino)acetate hydrochloride

075098-1G 075098-5G
102539-752EA 320 USD
102539-752 102539-754
Ethyl-2-((2-((tert-butoxycarbonyl)amino)ethyl)amino)acetate hydrochloride ≥95%
Ethyl-2-((2-((tert-butoxycarbonyl)amino)ethyl)amino)acetate hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₃ClN₂O₄
MDL Number: MFCD01862956
CAS Number: 347890-34-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR