4-Formylbenzoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: Terephthalaldehydic acid, 4-Carboxybenzaldehyde

075096-100G 075096-25G
102539-746EA 61.54 USD
102539-746 102833-776
4-Formylbenzoic acid ≥95%
4-Formylbenzoic acid

Hazard: Irritant/Air Sensitive/Stored under Argon

Formula: C₈H₆O₃
MW: 150.13 g/mol
Melting Pt: ca 245°C
MDL Number: MFCD00006951
CAS Number: 619-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR