1-Methyl-1H-pyrazole-4-boronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (1-Methyl-1H-pyrazol-4-yl)boronic acid

075094-1G 075094-5G
102539-734EA 92.31 USD
102539-734 102539-736
1-Methyl-1H-pyrazole-4-boronic acid ≥95%
1-Methyl-1H-pyrazole-4-boronic acid

Hazard: Irritant/Store in Freezer

Formula: C₄H₇BN₂O₂
MW: 125.92 g/mol
Melting Pt: 77…78 °C
MDL Number: MFCD09414709
CAS Number: 847818-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR