5-Chloro-2-phenyl-1H-indole ≥95%
Supplier: Matrix Scientific
075092-250MG
075092-1G
075092-5G
102539-722EA
200
USD
102539-722
102539-724
102539-726
5-Chloro-2-phenyl-1H-indole ≥95%
5-Chloro-2-phenyl-1H-indole
Hazard: Irritant
Formula:
C14H10ClN MW: 227.6889 g/mol |
MDL Number:
MFCD00034706 CAS Number: 23746-76-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...