5-Hydroxy-3H-benzimidazole ≥95%

Supplier: Matrix Scientific

Synonyms: 3H-Benzimidazol-5-ol, 1H-Benzimidazol-5-ol, 5-Hydroxybenzimidazole, 1H-Benzimidazol-6-ol, 1H-Benzo[d]imidazol-5-ol, 6-Hydroxy-1H-benzimidazole

075091-1G 075091-5G
102539-718EA 292.31 USD
102539-718 102539-720
5-Hydroxy-3H-benzimidazole ≥95%
5-Hydroxy-3H-benzimidazole

Hazard: Irritant

Formula: C₇H₆N₂O
MW: 134.14 g/mol
MDL Number: MFCD02241303
CAS Number: 41292-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR