Methyl-3,5-dibromobenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3,5-dibromobenzoate

075090-1G 075090-5G
102539-714EA 178.46 USD
102539-714 102539-716
Methyl-3,5-dibromobenzoate ≥95%
Methyl 3,5-dibromobenzoate

Hazard: Irritant

Formula: C₈H₆Br₂O₂
MDL Number: MFCD00082648
CAS Number: 51329-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR